3-[3-Ethyl-1-(4-isopropylphenyl)-1H-1,2,4-triazol-5-yl]propanoic acid

Supplier: Matrix Scientific

040527-500MG
101849-688EA 484.62 USD
101849-688
3-[3-Ethyl-1-(4-isopropylphenyl)-1H-1,2,4-triazol-5-yl]propanoic acid
3-[3-Ethyl-1-(4-isopropylphenyl)-1H-1,2,4-triazol-5-yl]propanoic acid

Hazard: Irritant

Formula: C₁₆H₂₁N₃O₂
MDL Number: MFCD12028237
CAS Number: 1142202-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR