2-Chloro-N-ethylpropanamide

Supplier: Matrix Scientific

040520-500MG
101849-674EA 290.77 USD
101849-674
2-Chloro-N-ethylpropanamide
2-Chloro-N-ethylpropanamide

Hazard: Irritant

Formula: C₅H₁₀ClNO
MDL Number: MFCD08239633
CAS Number: 67791-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR