2-Chloro-N-propylpropanamide

Supplier: Matrix Scientific

040517-500MG
101849-668EA 193.85 USD
101849-668
2-Chloro-N-propylpropanamide
2-Chloro-N-propylpropanamide

Hazard: Irritant

Formula: C₆H₁₂ClNO
MDL Number: MFCD09802012
CAS Number: 94318-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR