1-(2-Chloropropanoyl)pyrrolidine

Supplier: Matrix Scientific

040516-500MG
101849-666EA 290.77 USD
101849-666
1-(2-Chloropropanoyl)pyrrolidine
1-(2-Chloropropanoyl)pyrrolidine

Hazard: Irritant

Formula: C₇H₁₂ClNO
MDL Number: MFCD03986494
CAS Number: 75115-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR