1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
Supplier: Matrix Scientific
040513-500MG
101849-660EA
290.77
USD
101849-660
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-3-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₆N₂O₃ |
MDL Number:
MFCD03830326 CAS Number: 702670-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...