1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

040512-500MG
101849-658EA 193.85 USD
101849-658
1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid
1-(Dimethylcarbamoyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₆N₂O₃
MDL Number: MFCD03830325
CAS Number: 333985-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR