N-Propionylglycine

Supplier: Matrix Scientific

040509-500MG
101849-652EA 290.77 USD
101849-652
N-Propionylglycine
N-Propionylglycine

Hazard: Irritant

Formula: C₅H₉NO₃
MDL Number: MFCD09049674
CAS Number: 21709-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR