1-(Ethoxyacetyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

040507-500MG
101849-648EA 193.85 USD
101849-648
1-(Ethoxyacetyl)piperidine-4-carboxylic acid
1-(Ethoxyacetyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD11053940
CAS Number: 1042796-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR