1-(Methoxyacetyl)piperidine-3-carboxylic acid

Supplier: Matrix Scientific

040506-500MG
101849-646EA 290.77 USD
101849-646
1-(Methoxyacetyl)piperidine-3-carboxylic acid
1-(Methoxyacetyl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD09048090
CAS Number: 926238-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR