1-(Methoxyacetyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

040505-500MG
101849-644EA 290.77 USD
101849-644
1-(Methoxyacetyl)piperidine-4-carboxylic acid
1-(Methoxyacetyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD09046374
CAS Number: 926189-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR