1-Acetylpiperidine-3-carboxylic acid

Supplier: Matrix Scientific

040504-500MG
101849-642EA 320 USD
101849-642
1-Acetylpiperidine-3-carboxylic acid
1-Acetylpiperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD07348751
CAS Number: 2637-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR