[(1,1-Dimethylpropyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040497-500MG
101849-630EA 146.15 USD
101849-630
[(1,1-Dimethylpropyl)amino](oxo)acetic acid
[(1,1-Dimethylpropyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₇H₁₃NO₃
MDL Number: MFCD09971299
CAS Number: 1015846-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR