[(5-Methyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040495-500MG
101849-626EA 96.92 USD
101849-626
[(5-Methyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid
[(5-Methyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₅H₅N₃O₃S
MDL Number: MFCD09864507
CAS Number: 83244-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR