Oxo(piperidin-1-yl)acetic acid

Supplier: Matrix Scientific

040494-500MG
101849-624EA 146.15 USD
101849-624
Oxo(piperidin-1-yl)acetic acid
Oxo(piperidin-1-yl)acetic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD06740188
CAS Number: 4706-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR