[(2-Methylphenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040493-500MG
101849-622EA 146.15 USD
101849-622
[(2-Methylphenyl)amino](oxo)acetic acid
[(2-Methylphenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₉H₉NO₃
MDL Number: MFCD01001244
CAS Number: 406190-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR