(sec-Butylamino)(oxo)acetic acid

Supplier: Matrix Scientific

040491-500MG
101849-618EA 193.85 USD
101849-618
(sec-Butylamino)(oxo)acetic acid
(sec-Butylamino)(oxo)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD09971254
CAS Number: 80638-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR