3,4-Dihydroquinolin-1(2H)-yl(oxo)acetic acid
Supplier: Matrix Scientific
040490-500MG
101849-616EA
146.15
USD
101849-616
3,4-Dihydroquinolin-1(2H)-yl(oxo)acetic acid
3,4-Dihydroquinolin-1(2H)-yl(oxo)acetic acid
Hazard: Irritant
Formula:
C₁₁H₁₁NO₃ |
MDL Number:
MFCD10018451 CAS Number: 1018565-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...