[(2,4-Difluorophenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040488-500MG
101849-612EA 146.15 USD
101849-612
[(2,4-Difluorophenyl)amino](oxo)acetic acid
[(2,4-Difluorophenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₈H₅F₂NO₃
MDL Number: MFCD04047027
CAS Number: 678556-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR