[(2-Chlorophenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040487-500MG
101849-610EA 146.15 USD
101849-610
[(2-Chlorophenyl)amino](oxo)acetic acid
[(2-Chlorophenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₈H₆ClNO₃
MDL Number: MFCD10018463
CAS Number: 77901-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR