(Isobutylamino)(oxo)acetic acid

Supplier: Matrix Scientific

040485-500MG
101849-606EA 153.85 USD
101849-606
(Isobutylamino)(oxo)acetic acid
(Isobutylamino)(oxo)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD01418401
CAS Number: 75235-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR