Azepan-1-yl(oxo)acetic acid

Supplier: Matrix Scientific

040484-500MG
101849-604EA 193.85 USD
101849-604
Azepan-1-yl(oxo)acetic acid
Azepan-1-yl(oxo)acetic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD06740189
CAS Number: 886505-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR