2,3-Dihydro-1H-indol-1-yl(oxo)acetic acid

Supplier: Matrix Scientific

040482-500MG
101849-600EA 146.15 USD
101849-600
2,3-Dihydro-1H-indol-1-yl(oxo)acetic acid
2,3-Dihydro-1H-indol-1-yl(oxo)acetic acid

Hazard: Irritant

Formula: C₁₀H₉NO₃
MDL Number: MFCD10018450
CAS Number: 1018243-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR