[(4-Methylphenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-Oxo-2-(p-tolylamino)acetic acid

040481-500MG
101849-598EA 146.15 USD
101849-598
[(4-Methylphenyl)amino](oxo)acetic acid
[(4-Methylphenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD00209417
CAS Number: 42868-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR