(2-Methylpiperidin-1-yl)(oxo)acetic acid

Supplier: Matrix Scientific

040480-500MG
101849-596EA 153.85 USD
101849-596
(2-Methylpiperidin-1-yl)(oxo)acetic acid
(2-Methylpiperidin-1-yl)(oxo)acetic acid

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD09971294
CAS Number: 77654-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR