Oxo(propylamino)acetic acid

Supplier: Matrix Scientific

040477-500MG
101849-590EA 193.85 USD
101849-590
Oxo(propylamino)acetic acid
Oxo(propylamino)acetic acid

Hazard: Irritant

Formula: C₅H₉NO₃
MDL Number: MFCD09971257
CAS Number: 81682-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR