Azocan-1-yl(oxo)acetic acid
Supplier: Matrix Scientific
040475-500MG
101849-586EA
193.85
USD
101849-586
Azocan-1-yl(oxo)acetic acid
Azocan-1-yl(oxo)acetic acid
Hazard: Irritant
Formula:
C₉H₁₅NO₃ |
MDL Number:
MFCD12028233 CAS Number: 1142202-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...