[Methyl(phenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(Methyl(phenyl)amino)-2-oxoacetic acid

040473-500MG
101849-582EA 138.46 USD
101849-582
[Methyl(phenyl)amino](oxo)acetic acid
[Methyl(phenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₉H₉NO₃
MW: 179.18 g/mol
MDL Number: MFCD09864583
CAS Number: 13120-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR