[(3-Fluorophenyl)amino](oxo)acetic acid

Supplier: Matrix Scientific

040472-500MG
101849-580EA 146.15 USD
101849-580
[(3-Fluorophenyl)amino](oxo)acetic acid
[(3-Fluorophenyl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₈H₆FNO₃
MDL Number: MFCD09971235
CAS Number: 87967-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR