2-Anilino-2-oxoacetic acid

Supplier: Matrix Scientific

Synonyms: 2-Oxo-2-(phenylamino)acetic acid

040468-500MG
101849-572EA 307.69 USD
101849-572
2-Anilino-2-oxoacetic acid
2-Anilino-2-oxoacetic acid

Hazard: Irritant

Formula: C₈H₇NO₃
MW: 165.15 g/mol
MDL Number: MFCD00020384
CAS Number: 500-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR