(Cyclopentylamino)(oxo)acetic acid

Supplier: Matrix Scientific

040467-500MG
101849-570EA 290.77 USD
101849-570
(Cyclopentylamino)(oxo)acetic acid
(Cyclopentylamino)(oxo)acetic acid

Hazard: Irritant

Formula: C₇H₁₁NO₃
MDL Number: MFCD10018379
CAS Number: 183235-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR