(tert-Butylamino)(oxo)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(tert-Butylamino)-2-oxoacetic acid

040463-500MG
101849-562EA 387.69 USD
101849-562
(tert-Butylamino)(oxo)acetic acid
(tert-Butylamino)(oxo)acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₃
MW: 145.16 g/mol
MDL Number: MFCD02625536
CAS Number: 169772-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR