N-(sec-Butyl)-2-chloroacetamide

Supplier: Matrix Scientific

040457-500MG
101849-552EA 307.69 USD
101849-552
N-(sec-Butyl)-2-chloroacetamide
N-(sec-Butyl)-2-chloroacetamide

Hazard: Irritant

Formula: C₆H₁₂ClNO
MDL Number: MFCD00791326
CAS Number: 32322-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR