3-Bromo-N-methylbenzamide

Supplier: Matrix Scientific

040454-500MG
101849-542EA 290.77 USD
101849-542
3-Bromo-N-methylbenzamide
3-Bromo-N-methylbenzamide

Hazard: Irritant

Formula: C₈H₈BrNO
MW: 214.0592 g/mol
MDL Number: MFCD01060879
CAS Number: 49834-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR