1-(2,6-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

040445-500MG
101849-524EA 230.77 USD
101849-524
1-(2,6-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(2,6-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD05096196
CAS Number: 63674-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR