1-(3,5-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

040443-500MG
101849-520EA 290.77 USD
101849-520
1-(3,5-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3,5-Dimethylphenyl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD01571215
CAS Number: 63674-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR