5-Oxo-1-(1,1,3,3-tetramethylbutyl)pyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

040440-500MG
101849-514EA 290.77 USD
101849-514
5-Oxo-1-(1,1,3,3-tetramethylbutyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(1,1,3,3-tetramethylbutyl)pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₃NO₃
MDL Number: MFCD12028230
CAS Number: 1142202-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR