1-[2-(5-Chloro-1H-indol-3-yl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

040435-500MG
101849-506EA 290.77 USD
101849-506
1-[2-(5-Chloro-1H-indol-3-yl)ethyl]-5-oxopyrrolidine-3-carboxylic acid
1-[2-(5-Chloro-1H-indol-3-yl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₃
MDL Number: MFCD09271355
CAS Number: 931374-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR