[(3,4-Dichlorobenzyl)sulfonyl]acetic acid

Supplier: Matrix Scientific

040430-500MG
101849-496EA 290.77 USD
101849-496
[(3,4-Dichlorobenzyl)sulfonyl]acetic acid
[(3,4-Dichlorobenzyl)sulfonyl]acetic acid

Hazard: Irritant

Formula: C₉H₈Cl₂O₄S
MDL Number: MFCD10691573
CAS Number: 300700-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR