[(3,4-Dichlorobenzyl)thio]acetic acid

Supplier: Matrix Scientific

040429-500MG
101849-494EA 115.38 USD
101849-494
[(3,4-Dichlorobenzyl)thio]acetic acid
[(3,4-Dichlorobenzyl)thio]acetic acid

Hazard: Irritant

Formula: C₉H₈Cl₂O₂S
MDL Number: MFCD02755446
CAS Number: 65051-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR