2-(Isopropylthio)benzoic acid

Supplier: Matrix Scientific

040426-500MG
101849-488EA 233.85 USD
101849-488
2-(Isopropylthio)benzoic acid
2-(Isopropylthio)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₂S
MW: 196.27 g/mol
MDL Number: MFCD07364761
CAS Number: 41394-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR