2-(Butylsulfonyl)benzoic acid

Supplier: Matrix Scientific

040424-500MG
101849-484EA 290.77 USD
101849-484
2-(Butylsulfonyl)benzoic acid
2-(Butylsulfonyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄S
MDL Number: MFCD09947080
CAS Number: 57076-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR