2-(Butylthio)benzoic acid

Supplier: Matrix Scientific

040423-500MG
101849-482EA 193.85 USD
101849-482
2-(Butylthio)benzoic acid
2-(Butylthio)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂S
MDL Number: MFCD07364775
CAS Number: 17839-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR