2-(Propylthio)benzoic acid

Supplier: Matrix Scientific

Synonyms: 2-(Propylsulfanyl)benzoic acid2-(Propylsulphanyl)benzoic acid

040422-500MG
101849-480EA 193.85 USD
101849-480
2-(Propylthio)benzoic acid
2-(Propylthio)benzoic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₂S
MW: 196.27 g/mol
MDL Number: MFCD02729229
CAS Number: 21213-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR