[(4-Nitrobenzyl)thio]acetic acid

Supplier: Matrix Scientific

040421-500MG
101849-478EA 193.85 USD
101849-478
[(4-Nitrobenzyl)thio]acetic acid
[(4-Nitrobenzyl)thio]acetic acid

Hazard: Irritant

Formula: C₉H₉NO₄S
MDL Number: MFCD00540019
CAS Number: 6345-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR