2-[(3-Methylbutyl)thio]benzoic acid
Supplier: Matrix Scientific
040420-500MG
101849-476EA
153.85
USD
101849-476
2-[(3-Methylbutyl)thio]benzoic acid
2-[(3-Methylbutyl)thio]benzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₆O₂S |
MDL Number:
MFCD07364786 CAS Number: 17839-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...