3-[(4-Chlorophenyl)sulfanyl]propanoic acid
Supplier: Matrix Scientific
040416-500MG
101849-468EA
193.85
USD
101849-468
3-[(4-Chlorophenyl)sulfanyl]propanoic acid
3-[(4-Chlorophenyl)sulfanyl]propanoic acid
Hazard: Irritant
Formula:
C₉H₉ClO₂S |
MDL Number:
MFCD02636303 CAS Number: 6310-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...