[(4-Chlorobenzyl)sulfonyl]acetic acid

Supplier: Matrix Scientific

040414-500MG
101849-464EA 255.38 USD
101849-464
[(4-Chlorobenzyl)sulfonyl]acetic acid
[(4-Chlorobenzyl)sulfonyl]acetic acid

Hazard: Irritant

Formula: C₉H₉ClO₄S
MDL Number: MFCD06208470
CAS Number: 118672-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR