(Phenylsulfonyl)acetic acid
Supplier: Matrix Scientific
040412-500MG
101849-460EA
290.77
USD
101849-460
(Phenylsulfonyl)acetic acid
(Phenylsulfonyl)acetic acid
Hazard: Irritant
Formula:
C₈H₈O₄S |
MDL Number:
MFCD00025042 CAS Number: 3959-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...