2-[(3-Phenylpropyl)thio]benzoic acid

Supplier: Matrix Scientific

040410-500MG
101849-456EA 193.85 USD
101849-456
2-[(3-Phenylpropyl)thio]benzoic acid
2-[(3-Phenylpropyl)thio]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₂S
MDL Number: MFCD09271353
CAS Number: 366801-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR