5-(4-Methylbenzyl)-2-furoic acid
Supplier: Matrix Scientific
040403-500MG
101849-442EA
255.38
USD
101849-442
5-(4-Methylbenzyl)-2-furoic acid
5-(4-Methylbenzyl)-2-furoic acid
Hazard: Irritant
Formula:
C₁₃H₁₂O₃ |
MDL Number:
MFCD02320581 CAS Number: 4664-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...